n-Hexacosane 99%
Glass bottle for solids, 1 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: N-hexacosane
- CAS Number:630-01-3
- Formula:H₃C(CH₂)₂₄CH₃
- MDL Number:MFCD00009354
- Molecular Weight:366.71 g/mol
- Boiling Pt:420 °C
- Melting Pt:56…59 °C
- Density:0.8 g/cm³
- Flash Pt:215 °C
- Storage Temperature:Ambient
- EINECS:211-124-1
Documents
ACRO22122
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline solid
- Infrared spectrum:Conforms
- Melting point:56 to 59 °C
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



