D-(-)-Sorbitol 97.0-100.5%, GPR RECTAPUR®
Plastic bottle for solids, 2,5 kg
Supplier: VWR Chemicals
D-(-)-Sorbitol 97.0-100.5%, GPR RECTAPUR®
Catalog # 302425B
Supplier: VWR Chemicals
CAS Number: 50-70-4
Jump to:
Product Details & Documents
Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
- CAS Number:50-70-4
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00004708
- Molecular Weight:182.17 g/mol
- Merck Index:13,08797
- Boiling Pt:295 °C (1013 hPa)
- Melting Pt:94…98 °C
- Density:1.524 g/cm³ (20 °C)
- Flash Pt:>100 °C
- Storage Temperature:Ambient
- EINECS:200-061-5
Documents
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 6L Plastic bottle for solids DIN 80 (ZPACK13256000)
Packaging Information
Specifications
Specification Test Results
- Assay (calculated on anhydrous):97.0- 100.5 %
- Appearance of solution (10 %; water):Passes test
- Identification:Passes test
- Spec. opt. rot.(10 %; borate; anhydrous):4.0 - 7.0 °
- Reducing sugars (calculated as C6H12O6):Max. 0.2 %
- Ignition residue (SO4):Max. 0.1 %
- Water:Max. 1.5 %
- Cl (Chloride):Max. 50 ppm
- SO4 (Sulphate):Max. 100 ppm
- Ni (Nickel):Max. 1 ppm
- Pb (Lead):Max. 0.5 ppm
Specifications
- Pack type:Plastic bottle for solids
- Pk:2,5 kg



