Lead(IV) oxide 97+%, extra pure
Glass bottle for solids, 500 g
: Lead dioxide
- :1309-60-0
- :O₂Pb
- :MFCD00011165
- :239.2 g/mol
- :12,05424
- :290 °C (dec.)
- :9,4 g/cm³ (20 °C)
- :Ambient
- :215-174-5
- :1872
- :5.1,III
- Appearance:Brown to black Powder
- Titration with Na2S2O3:≥97.0 %
- Chloride (Cl):≤0.02 %
- Sulfate (SO4):≤0.005 %
- Copper (Cu):≤0.01 %
- Iron (Fe):≤0.01 %
- Zinc (Zn):≤0.005 %
- Pack type:Glass bottle for solids
- Pk:500 g



