4'-Ethoxyacetanilide 97%
Plastic bottle, 500 g
Supplier: Thermo Fisher Scientific
4'-Ethoxyacetanilide 97%
Catalog # ACRO216595000
Supplier: Thermo Fisher Scientific
CAS Number: 62-44-2
Jump to:
Product Details & Documents
Synonyms: Acetphenetidin, Acetophenetidin, Phenacetin, N-(4-Ethoxyphenyl)acetamide, 4-Acetophenetidide
- CAS Number:62-44-2
- Formula:C₁₀H₁₃NO₂
- MDL Number:MFCD00009094
- Molecular Weight:179.22 g/mol
- Melting Pt:133…138 °C
- Storage Temperature:Ambient
- EINECS:200-533-0
Documents
ACRO21659
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Melting point:133 to 138 °C
- HPLC:≥96.0 %
Specifications
- Pack type:Plastic bottle
- Pk:500 g



