Order Entry
Export
Orders LinkContactUsLinkComponent
L-thyroxine 97+%

L-thyroxine 97+%

Glass bottle, 1 g
Catalog # ACRO215660010
L-thyroxine 97+%
Catalog # ACRO215660010
CAS Number:  51-48-9
L-thyroxine 97+%
L-thyroxine 97+%

Glass bottle, 1 g

CAS Number:  51-48-9
Price
174,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  (S)-2-Amino-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid
  • CAS Number:51-48-9
  • Formula:C₁₅H₁₁I₄NO₄
  • MDL Number:MFCD00002595
  • Molecular Weight:776.874 g/mol
  • Melting Pt:235…236 °C (dec.)
  • Storage Temperature:Refrigerator

Documents

Specifications

Specification Test Results

  • Appearance:
    White to beige Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1.0 % (110°C, 4 h)
  • HPLC:
    ≥97.0 %
  • Specific optical rotation:
    -4.0° to -6.0° (20°C, 589 nm) (c=1,1N NaOH:EtOH 1:2) (on dried substance)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g

Specifications