Tungsten trioxide 99+%
Plastic bottle, 100 g
: Tungsten (VI) oxide
- :1314-35-8
- :O₃W
- :MFCD00011466
- :231.84 g/mol
- :12,09947
- :1470…1475 °C
- :7,16 g/cm³ (20 °C)
- :Density: 7.16 g/cm³ (20 °C)
- :Ambient
- :215-231-4
- Appearance:Yellow to yellow-green Powder
- Assay:≥99.0 %
- Pack type:Plastic bottle
- Pk:100 g



