Order Entry
Export
Orders LinkContactUsLinkComponent
Molybdic acid for analysis

Molybdic acid for analysis

Plastic bottle, 100 g
Catalog # ACRO213621000
Irritant
Health
Molybdic acid for analysis
Catalog # ACRO213621000
CAS Number:  7782-91-4
Molybdic acid for analysis
Molybdic acid for analysis

Plastic bottle, 100 g

CAS Number:  7782-91-4
Price
68,90 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:7782-91-4
  • Formula:H₂MoO₄
  • MDL Number:MFCD00036287
  • Molecular Weight:161.95 g/mol
  • Merck Index:13,06262
  • Boiling Pt:∼300 °C (1013 hPa)
  • Melting Pt:300 °C
  • Density:3.1 g/cm³
  • Storage Temperature:Ambient
  • EINECS:231-970-5

Documents

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Powder
  • Additional info:
    contains ammonium molybdate
  • Titration with NaOH:
    ≥85 % (as MoO3)
  • Chloride (Cl):
    ≤100 ppm
  • Phosphate (PO4):
    ≤5 ppm
  • Sulfate (SO4):
    ≤2000 ppm
  • Iron (Fe):
    ≤50 ppm
  • Heavy metals (as Pb):
    ≤30 ppm

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g

Specifications