Cetyldimethylethylammonium bromide 99%
Glass bottle, 500 g
Supplier: Thermo Fisher Scientific
Cetyldimethylethylammonium bromide 99%
Catalog # ACRO211875000
Supplier: Thermo Fisher Scientific
CAS Number: 124-03-8
Jump to:
Product Details & Documents
Synonyms: Ethylhexadecyldimethylammonium Bromide, Hexadecylethyldimethylammonium bromide, Ethyl(1-hexadecyl)dimethylammonium bromide, Ethylhexadecyldimethylammonium bromide, Dimethylethylhexadecylammonium bromide
- CAS Number:124-03-8
- Formula:C₂₀H₄₄BrN
- MDL Number:MFCD00011774
- Molecular Weight:378.4741 g/mol
- Melting Pt:190 °C (dec.)
- Storage Temperature:Ambient
- EINECS:204-672-8
- ADR:6.1,III
Documents
ACRO21187
SDS
Specifications
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:500 g



