2-Propylvaleric acid 99%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
2-Propylvaleric acid 99%
Catalog # ACRO208331000
Supplier: Thermo Fisher Scientific
CAS Number: 99-66-1
Jump to:
Product Details & Documents
Synonyms: 2-Propylpentanoic acid, Valproic Acid, 2,2-Di-n-propylacetic acid
- CAS Number:99-66-1
- Formula:C₈H₁₆O₂
- MDL Number:MFCD00002672
- Molecular Weight:144.21 g/mol
- Merck Index:12,10049
- Boiling Pt:220 °C (1013 hPa)
- Density:0.92 g/cm³
- Flash Pt:111 °C
- Storage Temperature:Ambient
- EINECS:202-777-3
- UN:3265
- ADR:8,III
Documents
ACRO20833
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4240 to 1.4260 (20°C, 589 nm)
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



