Order Entry
Export
Orders LinkContactUsLinkComponent
Sodium peroxide 96%

Sodium peroxide 96%

Plastic bottle, 25 g
Catalog # ACRO207700250
Corrosive
Oxidizing
Sodium peroxide 96%
Catalog # ACRO207700250
CAS Number:  1313-60-6
Sodium peroxide 96%
Sodium peroxide 96%

Plastic bottle, 25 g

CAS Number:  1313-60-6
Price
41,70 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  di-Sodium peroxide
  • CAS Number:1313-60-6
  • Formula:Na₂O₂
  • MDL Number:MFCD00003497
  • Molecular Weight:77.98 g/mol
  • Merck Index:13,08728
  • Melting Pt:460 °C (dec.)
  • Density:2.8 g/cm³
  • Storage Temperature:Ambient
  • EINECS:215-209-4
  • UN:1504
  • ADR:5.1,I

Documents

Specifications

Specification Test Results

  • Appearance:
    Yellow Granules (May contain a small quantity of discolored material from the manufacturing process)
  • Titration with KMnO4:
    ≥95 %
  • Chloride (Cl):
    ≤50 ppm
  • Sulfate (SO4):
    ≤50 ppm
  • Iron (Fe):
    ≤50 ppm
  • Lead (Pb):
    ≤50 ppm

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 g

Specifications