Sodium nitrate 99+% for analysis
Plastic bottle, 1 kg
Supplier: Thermo Fisher Scientific
Sodium nitrate 99+% for analysis
Catalog # ACRO205960010
Supplier: Thermo Fisher Scientific
CAS Number: 7631-99-4
Jump to:
Product Details & Documents
- CAS Number:7631-99-4
- Formula:NaNO₃
- MDL Number:MFCD00011119
- Molecular Weight:84.99 g/mol
- Merck Index:13,08720
- Boiling Pt:380 °C (1013 hPa)
- Melting Pt:306 °C
- Density:2,26 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-554-3
- UN:1498
- ADR:5.1,III
- REACH:01-2119488221-41
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals
- pH:5.5 to 8.3 (5 % at 25°C)
- Titration after Ion exchange:≥99.0 %
- Chloride (Cl):≤10 ppm
- Nitrite (NO2):≤10 ppm
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤30 ppm
- Calcium (Ca):≤50 ppm
- Iron (Fe):≤3 ppm
- Heavy metals (as Pb):≤5 ppm
Specifications
- Pack type:Plastic bottle
- Pk:1 kg



