cis-4-Hydroxy-D(+)-proline 99%
Glass bottle for solids, 1 g
Supplier: Thermo Fisher Scientific
cis-4-Hydroxy-D(+)-proline 99%
Catalog # ACRO204910010
Supplier: Thermo Fisher Scientific
CAS Number: 2584-71-6
Jump to:
Product Details & Documents
Synonyms: (R)-(+)-cis-4-Hydroxyproline, cis-4-Hydroxy-D-proline, H-D-cis-Hyp-OH, H-cis-D-Hyp-OH, D(+)-cis-4-Hydroxyproline
- CAS Number:2584-71-6
- Formula:C₅H₉NO₃
- MDL Number:MFCD00005252
- Molecular Weight:131.13 g/mol
- Melting Pt:243 °C (dec.)
- Storage Temperature:Ambient
- EINECS:219-963-5
Documents
ACRO20491
SDS
Specifications
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- TLC:≥98.5 %
- Specific optical rotation:+56° to +60° (24°C, 589 nm) (c=2, water)
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



