Hexamethyldisilane 98+%
Glass bottle, 10 mL
Supplier: Thermo Fisher Scientific
Hexamethyldisilane 98+%
Catalog # ACRO202690100
Supplier: Thermo Fisher Scientific
CAS Number: 1450-14-2
Jump to:
Product Details & Documents
- CAS Number:1450-14-2
- Formula:C₆H₁₈Si₂
- MDL Number:MFCD00008258
- Molecular Weight:146.38 g/mol
- Boiling Pt:112 °C (1013 hPa)
- Melting Pt:15 °C
- Density:0.71 g/cm³
- Flash Pt:11 °C
- Storage Temperature:Ambient
- EINECS:215-911-0
- UN:1993
- ADR:3,II
Documents
ACRO20269
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4211 to 1.4231 (20°C, 589 nm)
- GC:≥98.0 %
Specifications
- Pack type:Glass bottle
- Pk:10 mL



