Orders LinkContactUsLinkComponent
Pressure Bulbs
Pressure Bulbs
Atomiser bulb with pressure valve, red-brown, size 3, PVC, 65
Catalog # 612-9952
Supplier:   DEUTSCH NEUMANN
Pressure Bulbs
Catalog # 612-9952
Supplier:  DEUTSCH NEUMANN
Pressure Bulbs
Pressure Bulbs
Atomiser bulb with pressure valve, red-brown, size 3, PVC, 65
Price
23,30 €
Log in for availability
1 * 1 items
Product Details & Documents
Natural rubber, chloroprene or PVC.
Used for filling automatic burettes, Pellet pattern (ball set with net) or atomisers (bulb).
Specifications
  • Capacity:
    65
  • Description:
    Atomiser bulb with pressure valve, red-brown, size 3, PVC
  • Seq:
    1