(±)-1,2-Octanediol 98+%
Glass bottle for solids, 50 g
Supplier: Thermo Fisher Scientific
(±)-1,2-Octanediol 98+%
Catalog # ACRO200140500
Supplier: Thermo Fisher Scientific
CAS Number: 1117-86-8
Jump to:
Product Details & Documents
Synonyms: 1,2-Octanediol, (±)-1,2-Octylene glycol, (±)-1,2-Dihydroxy octane
- CAS Number:1117-86-8
- Formula:C₈H₁₈O₂
- MDL Number:MFCD00010738
- Molecular Weight:146.23 g/mol
- Boiling Pt:243 °C (1013 hPa)
- Melting Pt:28…31 °C
- Flash Pt:138 °C
- EINECS:214-254-7
Documents
Specifications
Specification Test Results
- Appearance:Colorless to white Low melting solid
- Infrared spectrum:Conforms
- GC:≥98.0 %
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



