Order Entry
Export
Orders LinkContactUsLinkComponent
Lead(II) oxide 99.7%

Lead(II) oxide 99.7%

Glass bottle, 1 kg
Catalog # ACRO199340010
Irritant
Health
Environment
Lead(II) oxide 99.7%
Catalog # ACRO199340010
CAS Number:  1317-36-8
Lead(II) oxide 99.7%
Lead(II) oxide 99.7%

Glass bottle, 1 kg

CAS Number:  1317-36-8
Price
70,60 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Lead monoxide
  • CAS Number:1317-36-8
  • Formula:PbO
  • MDL Number:MFCD00011164
  • Molecular Weight:223.2 g/mol
  • Merck Index:13,05431
  • Boiling Pt:1470 °C
  • Melting Pt:886 °C
  • Density:9,5 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-267-0
  • UN:3077
  • ADR:9,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Yellow Powder
  • Titration Complexometric:
    ≥99.6 %
  • Loss on drying:
    ≤0.1 % (105°C)
  • Copper (Cu):
    ≤10 ppm
  • Iron (Fe):
    ≤20 ppm
  • Lead (Pb):
    ≤0.01 %
  • Silicon (Si):
    ≤20 ppm
  • Lead oxide (Pb3O4):
    ≤0.02 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 kg

Specifications