Order Entry
Export
Orders LinkContactUsLinkComponent
 

Lead(II) iodide 99%

Glass bottle for solids, 250 g
Catalog # ACRO198862500
Irritant
Health
Environment
Lead(II) iodide 99%
Catalog # ACRO198862500
CAS Number:  10101-63-0
Lead(II) iodide 99%
Lead(II) iodide 99%

Glass bottle for solids, 250 g

CAS Number:  10101-63-0
Price
271,00 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Lead diiodide
  • CAS Number:10101-63-0
  • Formula:I₂Pb
  • MDL Number:MFCD00011163
  • Molecular Weight:461.01 g/mol
  • Merck Index:12,05430
  • Boiling Pt:954 °C (1013 hPa)
  • Melting Pt:402 °C
  • Density:6.16 g/cm³
  • EINECS:233-256-9
  • UN:2291
  • ADR:6.1,III

 

Specifications

Specification Test Results

  • Appearance:
    Yellow to orange Powder
  • Titration Argentometric:
    ≥98.5 %
  • Loss on drying:
    ≤0.1 %
  • Chloride (Cl):
    ≤100 ppm

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications