Lead(II) bromide 98+%, extra pure
Glass bottle for solids, 500 g
: Lead dibromide, Lead(II) bromide
- :10031-22-8
- :Br₂Pb
- :MFCD00011156
- :367.01 g/mol
- :12,05418
- :916 °C (1013 hPa)
- :373 °C
- :6.66 g/cm³
- :233-084-4
- :2291
- :6.1,III
- Appearance:White to off-white Crystalline powder
- Titration Argentometric:≥98.0 %
- Loss on drying:≤0.1 %
- Chloride (Cl):≤100 ppm
- Insoluble matter:≤0.005 % (in acid)
- Pack type:Glass bottle for solids
- Pk:500 g



