Lead(II) bromide 98+%, extra pure
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Lead(II) bromide 98+%, extra pure
Catalog # ACRO198851000
Supplier: Thermo Fisher Scientific
CAS Number: 10031-22-8
Jump to:
Product Details & Documents
Synonyms: Lead dibromide, Lead(II) bromide
- CAS Number:10031-22-8
- Formula:Br₂Pb
- MDL Number:MFCD00011156
- Molecular Weight:367.01 g/mol
- Merck Index:12,05418
- Boiling Pt:916 °C (1013 hPa)
- Melting Pt:373 °C
- Density:6.66 g/cm³
- EINECS:233-084-4
- UN:2291
- ADR:6.1,III
Documents
ACRO19885
SDS
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Titration Argentometric:≥98.0 %
- Loss on drying:≤0.1 %
- Chloride (Cl):≤100 ppm
- Insoluble matter:≤0.005 % (in acid)
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



