Order Entry
Export
Orders LinkContactUsLinkComponent
Dipotassium hexachloroosmate > 39% Os

Dipotassium hexachloroosmate > 39% Os

Glass bottle for solids, 1 g
Catalog # ACRO197650010
Corrosive
Toxic
Dipotassium hexachloroosmate > 39% Os
Catalog # ACRO197650010
CAS Number:  16871-60-6
Dipotassium hexachloroosmate > 39% Os
Dipotassium hexachloroosmate > 39% Os

Glass bottle for solids, 1 g

CAS Number:  16871-60-6
Price
230,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Potassium hexachloroosmate(IV)
  • CAS Number:16871-60-6
  • Formula:Cl₆K₂Os
  • MDL Number:MFCD00011371
  • Molecular Weight:481.14 g/mol
  • Melting Pt:600 °C
  • Density:3,42 g/cm³ (20 °C)
  •  :Density: 3.42 g/cm³ (20 °C)
  • EINECS:240-893-6
  • UN:3290

Documents

Specifications

Specification Test Results

  • Appearance:
    Bordeaux to purple Powder
  • Assay:
    ≥39 % (Os)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications