Order Entry
Export
Orders LinkContactUsLinkComponent
Fluorescamine, pure

Fluorescamine, pure

Glass bottle, 25 mg
Catalog # ACRO191670250
Irritant
Fluorescamine, pure
Catalog # ACRO191670250
CAS Number:  38183-12-9
Fluorescamine, pure
Fluorescamine, pure

Glass bottle, 25 mg

CAS Number:  38183-12-9
Price
56,10 €
Log in for availability
1 * 25 mg
Product Details & Documents

CAS No.: 38183-12-9

Synonyms:  Fluram, 4-Phenyl-3H,3'H-spiro[furan-2,1'-isobenzofuran]-3,3'-dione, 4-Phenylspiro[furan-2(3H),1'-phthalan]-3,3'-dione
  • CAS Number:38183-12-9
  • Formula:C₁₇H₁₀O₄
  • MDL Number:MFCD00005928
  • Molecular Weight:278.26 g/mol
  • Melting Pt:155…157 °C
  • Storage Temperature:Ambient

Documents

Specifications

Specification Test Results

  • Appearance:
    Light yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    155 to 157 °C
  • Specific extinction E (1%/1cm):
    ≥900 (235 nm, ether)
  • Suitability:
    fluorescence ind. for prim. amines and amino acids

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mg

Specifications