Chlorophenol red, pure indicator grade
Glass bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
Chlorophenol red, pure indicator grade
Catalog # ACRO190040100
Supplier: Thermo Fisher Scientific
CAS Number: 4430-20-0
Jump to:
Product Details & Documents
Synonyms: 3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide
- CAS Number:4430-20-0
- Formula:C₁₉H₁₂Cl₂O₅S
- MDL Number:MFCD00005877
- Molecular Weight:423.27 g/mol
- Melting Pt:261 °C
- Storage Temperature:Ambient
- EINECS:224-619-2
Documents
ACRO19004
SDS
Specifications
Specification Test Results
- Appearance:Green to red-brown to brown Powder
- Infrared spectrum:Conforms
- Loss on drying:≤8 % (110°C)
- Solubility:(0.1 % in ethanol) Clear brown-yellow to dark orange
- UV Lambda max:572 to 576 nm (in 0.1 N NaOH)
- Specific absorbance A (1%/1cm):≥1000 (at Lambda max) On dried substance
- Visual transition interval:from pH 4.8 (yellow) to pH 6.4 (purple)
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



