Sterile containers, Falcon®
220 ml, with lid, Individually wrapped
Supplier: Corning
Jump to:
Product Details & Documents
PP, sterile, with or without PE lid.
- Moulded-in graduations for easy measurement
- Inert and chemically resistant to commonly used laboratory reagents at room temperature
Convenient, single-use containers for collection, transport and storage of a wide variety of specimens.
Polyethylene lid is frosted for easy marking, and fits both container sizes.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- Packed:Individually wrapped
- Capacity:220 ml
- Description:with lid
- Seq:04