Order Entry
Export
Orders LinkContactUsLinkComponent
2,2-Dibromopropane 98%

2,2-Dibromopropane 98%

Glass bottle for solids, 50 g
Catalog # ACRO186060500
Irritant
2,2-Dibromopropane 98%
Catalog # ACRO186060500
CAS Number:  594-16-1
2,2-Dibromopropane 98%
2,2-Dibromopropane 98%

Glass bottle for solids, 50 g

CAS Number:  594-16-1
Price
520,00 €
Log in for availability
1 * 50 g
Product Details & Documents
  • CAS Number:594-16-1
  • Formula:H₃CBr₂CCH₃
  • MDL Number:MFCD00000119
  • Molecular Weight:201.89 g/mol
  • Boiling Pt:114 °C (740 mmHg)
  • Melting Pt:–20 °C
  • Density:1.78 g/cm³
  • Flash Pt:>110 °C
  • EINECS:209-828-9

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4950 to 1.4990 (20°C, 589 nm)
  • GC:
    ≥97.5 %
  • Residue after ignition:
    ≤0.1 %
  • Specific optical rotation:
    -66° to -69° (20°C, 589 nm) (1 % in EtOH)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications