2,3-Dihydroxybenzaldehyde 97%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
2,3-Dihydroxybenzaldehyde 97%
Catalog # ACRO183510250
Supplier: Thermo Fisher Scientific
CAS Number: 24677-78-9
Jump to:
Product Details & Documents
- CAS Number:24677-78-9
- Formula:C₇H₆O₃
- MDL Number:MFCD00003324
- Molecular Weight:138.12 g/mol
- Boiling Pt:235 °C (1013 hPa)
- Melting Pt:104…109 °C
- Storage Temperature:Ambient
- EINECS:246-398-1
Documents
ACRO18351
SDS
Specifications
Specification Test Results
- Appearance:Light yellow to green-beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:104 to 109 °C
- GC:≥96.0 %
- Titration with NaOH:≥96.0 %
- Residue after ignition:≤0.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



