2,3-Dihydroxybenzaldehyde 97%
Glass bottle for solids, 5 g
- :24677-78-9
- :C₇H₆O₃
- :MFCD00003324
- :138.12 g/mol
- :235 °C (1013 hPa)
- :104…109 °C
- :Ambient
- :246-398-1
Documents
ACRO18351
SDS
- Appearance:Light yellow to green-beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:104 to 109 °C
- GC:≥96.0 %
- Titration with NaOH:≥96.0 %
- Residue after ignition:≤0.5 %
- Pack type:Glass bottle for solids
- Pk:5 g



