Formamide 99.5+%, extra pure
Plastic drum, 25 L
: Carbamaldehyde
- :75-12-7
- :HCONH₂
- :MFCD00007941
- :45.04 g/mol
- :13,04261
- :210 °C (760 mmHg)
- :2…3 °C
- :1.133 g/cm³
- :175 °C
- :Ambient
- :200-842-0
Documents
ACRO18109
SDS
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4460 to 1.4480 (20°C, 589 nm)
- Water:≤0.1 % (Coulometric)
- GC:≥99.50 %
- Residue after evaporation:≤0.05 %
- Pack type:Plastic drum
- Pk:25 L



