1,10-Decanediol 99%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1,10-Dihydroxy decane
- CAS Number:112-47-0
- Formula:C₁₀H₂₂O₂
- MDL Number:MFCD00004749
- Molecular Weight:174.28 g/mol
- Merck Index:13,02869
- Boiling Pt:297 °C (760 mmHg)
- Melting Pt:71…75 °C
- Density:0,883 g/cm³ (80 °C)
- Flash Pt:152 °C
- Storage Temperature:Ambient
- EINECS:203-975-2
Documents
ACRO18104
SDS
Specifications
Specification Test Results
- Appearance:White Powder or flakes or pellets
- Infrared spectrum:Conforms
- Melting point:72 to 74 °C
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



