Order Entry
Export
Orders LinkContactUsLinkComponent
 

Barbituric acid 99+%

Plastic bottle, 100 g
Catalog # ACRO180921000
Barbituric acid 99+%
Catalog # ACRO180921000
CAS Number:  67-52-7
Barbituric acid 99+%
Barbituric acid 99+%

Plastic bottle, 100 g

CAS Number:  67-52-7
Price
47,70 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  N,N'-Malonylurea, 2,4,6-Trihydroxypyrimidine
  • CAS Number:67-52-7
  • Formula:C₄H₄N₂O₃
  • MDL Number:MFCD00006666
  • Molecular Weight:128.09 g/mol
  • Merck Index:13,00966
  • Boiling Pt:441 °C (1013 hPa)
  • Melting Pt:248…255 °C
  • Flash Pt:150 °C
  • Storage Temperature:Ambient
  • EINECS:200-658-0
Specifications

Specification Test Results

  • Appearance:
    Cream Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    248 to 255 °C
  • Loss on drying:
    ≤0.5 %
  • Sulfated ash:
    ≤0.2 %
  • Titration with NaOH:
    99.0 to 101.0 %
  • Heavy metals (as Pb):
    ≤20 ppm

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 g

Specifications