1-Tetradecanol 99%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 1-Tetradecyl alcohol, 1-Hydroxy tetradecane, Myristyl alcohol
- CAS Number:112-72-1
- Formula:H₃C(CH₂)₁₂CH₂OH
- MDL Number:MFCD00004757
- Molecular Weight:214.39 g/mol
- Merck Index:13,06361
- Boiling Pt:289 °C (760 mmHg)
- Melting Pt:37…40 °C
- Density:0,8 g/cm³ (60 °C)
- Flash Pt:145 °C
- Storage Temperature:Ambient
- EINECS:204-000-3
- UN:3077
- ADR:9,III
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals or flakes or pellets and/or chunks
- Infrared spectrum:Conforms
- Melting point:37 to 41 °C
- GC:≥98.5 %
- Titration with HClO4:97.5 to 102.5 % (on dry substance)
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



