Order Entry
Export
Orders LinkContactUsLinkComponent
 

Sodium alginate

Plastic bottle, 500 g
Catalog # ACRO177775000
Sodium alginate
Catalog # ACRO177775000
CAS Number:  9005-38-3
Sodium alginate
Sodium alginate

Plastic bottle, 500 g

CAS Number:  9005-38-3
Price
180,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Alginic acid sodium salt, Algin
  • CAS Number:9005-38-3
  • MDL Number:MFCD00081310
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Appearance:
    Off-white to yellow-brown Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤15.0 % (100 mg, 105°C, 4 hrs)
  • Sulfated ash:
    30.0 % to 36.0 % (on dried substance)
  • Viscosity:
    350 to 550 mPa.s (1 % at 20°C) (Brookfield LV)
  • pH:
    5.5 to 8.0 (1 % at 20°C)
  • Heavy metals (as Pb):
    ≤20 ppm

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g

Specifications