Methanol 99.9% for analysis
Glass bottle, 500 mL
Methanol 99.9% for analysis
176845000.
: 67-56-1
: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- :67-56-1
- :CH₃OH
- :MFCD00004595
- :32.04 g/mol
- :13,05984
- :64.7 °C (760 mmHg)
- :–98 °C
- :0.791 g/cm³
- :12 °C
- :Ambient
- :200-659-6
- :1230
- :3,II
- :01-2119433307-44
Documents
- Appearance:Clear liquid
- Water:≤0.05 % (K.F.)
- Solubility:(in water) Passes test
- GC:≥99.85 %
- Color scale:≤5 APHA
- Residue after evaporation:≤0.001 %
- Acidity/alkalinity:≤0.0005 meq/g
- Substance darkened by H2SO4:≤5 APHA
- Calcium (Ca):≤0.2 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.02 ppm
- Magnesium (Mg):≤0.05 ppm
- Potassium (K):≤0.5 ppm
- Sodium (Na):≤0.5 ppm
- Zinc (Zn):≤0.05 ppm
- Carbonyl compounds:≤0.001 %
- Ethanol:≤0.05 %
- 2-Propanol:≤0.005 %
- KMnO4-reducing substance:≤0.00025 %
- Total phosphorus (P):≤0.05 ppm
- Total silicon (Si):≤0.05 ppm
- Total sulfur (S):≤0.1 ppm
- Pack type:Glass bottle
- Pk:500 mL
- Cat. No.:176845000.
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health



