Methanol 99.9% for analysis
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Methanol 99.9% for analysis
Catalog # 176840025.
Supplier: Thermo Fisher Scientific
CAS Number: 67-56-1
Jump to:
Product Details & Documents
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- CAS Number:67-56-1
- Formula:CH₃OH
- MDL Number:MFCD00004595
- Molecular Weight:32.04 g/mol
- Merck Index:13,05984
- Boiling Pt:64.7 °C (760 mmHg)
- Melting Pt:–98 °C
- Density:0.791 g/cm³
- Flash Pt:12 °C
- Storage Temperature:Ambient
- EINECS:200-659-6
- UN:1230
- ADR:3,II
- REACH:01-2119433307-44
Documents
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.05 % (K.F.)
- Solubility:(in water) Passes test
- GC:≥99.85 %
- Color scale:≤5 APHA
- Residue after evaporation:≤0.001 %
- Acidity/alkalinity:≤0.0005 meq/g
- Substance darkened by H2SO4:≤5 APHA
- Calcium (Ca):≤0.2 ppm
- Copper (Cu):≤0.02 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.02 ppm
- Magnesium (Mg):≤0.05 ppm
- Potassium (K):≤0.5 ppm
- Sodium (Na):≤0.5 ppm
- Zinc (Zn):≤0.05 ppm
- Carbonyl compounds:≤0.001 %
- Ethanol:≤0.05 %
- 2-Propanol:≤0.005 %
- KMnO4-reducing substance:≤0.00025 %
- Total phosphorus (P):≤0.05 ppm
- Total silicon (Si):≤0.05 ppm
- Total sulfur (S):≤0.1 ppm
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Cat. No.:176840025.
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health



