Diethyl ether 99+% stabilised, extra pure
Metal drum, 5 L
Supplier: Thermo Fisher Scientific
Diethyl ether 99+% stabilised, extra pure
Catalog # ACRO176830050
Supplier: Thermo Fisher Scientific
CAS Number: 60-29-7
Jump to:
Product Details & Documents
Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
- CAS Number:60-29-7
- Formula:(CH₃CH₂)₂O
- MDL Number:MFCD00011646
- Molecular Weight:74.12 g/mol
- Boiling Pt:34.6 °C (760 mmHg)
- Melting Pt:–116 °C
- Density:0.714 g/cm³
- Flash Pt:–45 °C
- Storage Temperature:Ambient
- UN:1155
- ADR:3,I
- :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3510 to 1.3540 (20°C, 589 nm)
- Water:≤0.1 % (Coulometric)
- GC:≥99.0 %
- Color scale:≤10 APHA
- Stabilizer:5 to 7 ppm BHT
- Relative density:(20°C) 0.714 to 0.716
- Residue after evaporation:≤0.005 %
- Acidity:≤0.001 meq/g
- Peroxides:≤0.0005 %
Specifications
- Pack type:Metal drum
- Pk:5 L



