Order Entry
Export
Orders LinkContactUsLinkComponent
Diethyl ether 99+% stabilised, extra pure

Diethyl ether 99+% stabilised, extra pure

Glass bottle, 1 L
Catalog # ACRO176830010
Irritant
Flammable
Diethyl ether 99+% stabilised, extra pure
Catalog # ACRO176830010
CAS Number:  60-29-7
Diethyl ether 99+% stabilised, extra pure
Diethyl ether 99+% stabilised, extra pure

Glass bottle, 1 L

CAS Number:  60-29-7
Price
51,70 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
  • CAS Number:60-29-7
  • Formula:(CH₃CH₂)₂O
  • MDL Number:MFCD00011646
  • Molecular Weight:74.12 g/mol
  • Boiling Pt:34.6 °C (760 mmHg)
  • Melting Pt:–116 °C
  • Density:0.714 g/cm³
  • Flash Pt:–45 °C
  • Storage Temperature:Ambient
  • UN:1155
  • ADR:3,I
  •  :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3510 to 1.3540 (20°C, 589 nm)
  • Water:
    ≤0.1 % (Coulometric)
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA
  • Stabilizer:
    5 to 7 ppm BHT
  • Relative density:
    (20°C) 0.714 to 0.716
  • Residue after evaporation:
    ≤0.005 %
  • Acidity:
    ≤0.001 meq/g
  • Peroxides:
    ≤0.0005 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications