Methyl propiolate 99%
Glass bottle for solids, 50 g
: Methyl acetylenecarboxylate, Propiolic acid methyl ester, 2-Propynoic acid methyl ester
- :922-67-8
- :C₄H₄O₂
- :MFCD00008572
- :84.07 g/mol
- :105 °C (1013 hPa)
- :0.94 g/cm³
- :10 °C
- :Refrigerator
- :213-083-5
- :3272
- :3,II
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4070 to 1.4090 (20°C, 589 nm)
- GC:≥98.5 %
- Chloride (Cl):8.6 to 9.6 %
- Pack type:Glass bottle for solids
- Pk:50 g



