Methyl propiolate 99%
Glass bottle for solids, 10 g
Supplier: Thermo Fisher Scientific
Methyl propiolate 99%
Catalog # ACRO174040100
Supplier: Thermo Fisher Scientific
CAS Number: 922-67-8
Jump to:
Product Details & Documents
Synonyms: Methyl acetylenecarboxylate, Propiolic acid methyl ester, 2-Propynoic acid methyl ester
- CAS Number:922-67-8
- Formula:C₄H₄O₂
- MDL Number:MFCD00008572
- Molecular Weight:84.07 g/mol
- Boiling Pt:105 °C (1013 hPa)
- Density:0.94 g/cm³
- Flash Pt:10 °C
- Storage Temperature:Refrigerator
- EINECS:213-083-5
- UN:3272
- ADR:3,II
Documents
ACRO17404
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4070 to 1.4090 (20°C, 589 nm)
- GC:≥98.5 %
- Chloride (Cl):8.6 to 9.6 %
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



