1,1-Dimethoxyacetone 98%
Glass bottle for solids, 500 g
: Pyruvaldehyde 1,1-dimethyl acetal, 1,1-Dimethoxypropan-2-one, Pyruvic aldehyde dimethyl acetal
- :6342-56-9
- :C₅H₁₀O₃
- :MFCD00008758
- :118.13 g/mol
- :143 °C (1013 hPa)
- :–57 °C
- :1 g/cm³
- :37 °C
- :228-735-4
- :1224
- :3,III
Documents
ACRO17380
SDS
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3970 to 1.3990 (20°C, 589 nm)
- GC:≥97.5 %
- Pack type:Glass bottle for solids
- Pk:500 g



