1,1-Dimethoxyacetone 98%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
1,1-Dimethoxyacetone 98%
Catalog # ACRO173801000
Supplier: Thermo Fisher Scientific
CAS Number: 6342-56-9
Jump to:
Product Details & Documents
Synonyms: Pyruvaldehyde 1,1-dimethyl acetal, 1,1-Dimethoxypropan-2-one, Pyruvic aldehyde dimethyl acetal
- CAS Number:6342-56-9
- Formula:C₅H₁₀O₃
- MDL Number:MFCD00008758
- Molecular Weight:118.13 g/mol
- Boiling Pt:143 °C (1013 hPa)
- Melting Pt:–57 °C
- Density:1 g/cm³
- Flash Pt:37 °C
- EINECS:228-735-4
- UN:1224
- ADR:3,III
Documents
ACRO17380
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3970 to 1.3990 (20°C, 589 nm)
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



