1,3-Dichloroacetone 99%
Glass bottle for solids, 50 g
Supplier: Thermo Fisher Scientific
1,3-Dichloroacetone 99%
Catalog # ACRO173630500
Supplier: Thermo Fisher Scientific
CAS Number: 534-07-6
Jump to:
Product Details & Documents
- CAS Number:534-07-6
- Formula:ClH₂CC(O)CH₂Cl
- MDL Number:MFCD00000937
- Molecular Weight:126.97 g/mol
- Merck Index:13,03077
- Boiling Pt:173 °C (1013 hPa)
- Melting Pt:38…42 °C
- Density:1.383 g/cm³
- Flash Pt:89 °C
- Storage Temperature:Refrigerator
- EINECS:208-585-6
- UN:2649
- ADR:6.1,II
Documents
ACRO17363
SDS
Specifications
Specification Test Results
- Appearance:Colorless to white Crystals
- Infrared spectrum:Conforms
- Melting point:38 to 42 °C
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



