Order Entry
Export
Orders LinkContactUsLinkComponent
D-Alanine 99+%

D-Alanine 99+%

Glass bottle for solids, 25 g
Catalog # ACRO172080250
D-Alanine 99+%
Catalog # ACRO172080250
CAS Number:  338-69-2
D-Alanine 99+%
D-Alanine 99+%

Glass bottle for solids, 25 g

CAS Number:  338-69-2
Price
193,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  (R)-(-)-Alanine, Alanine, D-α-Alanine, D(-)-Alanine, H-D-Ala-OH, α-Alanine
  • CAS Number:338-69-2
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00008077
  • Molecular Weight:89.09 g/mol
  • Merck Index:13,00203
  • Boiling Pt:189 °C (1013 hPa)
  • Melting Pt:278…282 °C (dec.)
  • Storage Temperature:Ambient
  • EINECS:206-418-1

Documents

Specifications

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.2 % (105°C, 3 hrs)
  • Sulfated ash:
    ≤0.2 %
  • HPLC:
    ≤1 % (L-content)
  • UV transmittance:
    ≥95 % (c=10, H2O, 430 nm) 1 cm cell
  • Specific optical rotation:
    -13.5° to -15.5° (20°C, 589 nm) (c=10, 6 N HCl) (on dry substance)
  • Titration with HClO4:
    99.0 to 101.0 % (On dry substance)
  • Chloride (Cl):
    ≤0.1 %
  • Arsenic (As):
    ≤2 ppm (As2O3)
  • Heavy metals (as Pb):
    ≤10 ppm

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications