Charcoal activated, NORIT® SA 2, decolorizing
Plastic drum, 1 kg
Supplier: Thermo Fisher Scientific
Charcoal activated, NORIT® SA 2, decolorizing
Catalog # ACRO171530010
Supplier: Thermo Fisher Scientific
CAS Number: 7440-44-0
Jump to:
Product Details & Documents
Synonyms: Carbon activated, Activated carbon, Activated charcoal
- CAS Number:7440-44-0
- Formula:C
- MDL Number:MFCD00133992
- Molecular Weight:12.01 g/mol
- Merck Index:13,01818
- Melting Pt:3652 °C
- Density:2 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-153-3
- ADR:4.2,III
- REACH:01-2119488894-16
Documents
ACRO17153
SDS
Specifications
Specification Test Results
- Appearance:Black Powder
- Loss on drying:≤10 %
- Molasses number:≤390
- Water:≤1.5 %
- GC:≥96.0 %
Specifications
- Pack type:Plastic drum
- Pk:1 kg



