Order Entry
Export
Orders LinkContactUsLinkComponent
Uric acid 99+%

Uric acid 99+%

Plastic bottle, 25 g
Catalog # ACRO171290250
Uric acid 99+%
Catalog # ACRO171290250
CAS Number:  69-93-2
Uric acid 99+%
Uric acid 99+%

Plastic bottle, 25 g

CAS Number:  69-93-2
Price
58,40 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1H-Purine-2,6,8(3H,7H,9H)-trione
  • CAS Number:69-93-2
  • Formula:C₅H₄N₄O₃
  • MDL Number:MFCD00005712
  • Molecular Weight:168.11 g/mol
  • Merck Index:12,10014
  • Melting Pt:>300 °C
  • Density:1,89 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-720-7

Documents

Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Assay:
    ≥99.0 % (ex N)
  • Loss on drying:
    ≤0.5 % (105°C, 3 hrs)
  • Sulfated ash:
    ≤0.2 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 g

Specifications