Order Entry
Export
Orders LinkContactUsLinkComponent
Quinaldic acid 99%

Quinaldic acid 99%

Glass bottle for solids, 10 g
Catalog # ACRO171050100
Irritant
Quinaldic acid 99%
Catalog # ACRO171050100
CAS Number:  93-10-7
Quinaldic acid 99%
Quinaldic acid 99%

Glass bottle for solids, 10 g

CAS Number:  93-10-7
Price
81,30 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Quinoline-2-carboxylic acid, 2-Quinolinecarboxylic acid
  • CAS Number:93-10-7
  • Formula:C₁₀H₇NO₂
  • MDL Number:MFCD00006752
  • Molecular Weight:173.17 g/mol
  • Merck Index:12,08227
  • Melting Pt:159…161 °C
  • Storage Temperature:Ambient
  • EINECS:202-218-3

Documents

Specifications

Specification Test Results

  • Appearance:
    Light brown Crystalline powder or needles
  • Infrared spectrum:
    Conforms
  • Melting point:
    157 to 159 °C
  • Titration with NaOH:
    ≥98.5 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications