Cyclohexanecarbonyl chloride 98%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Cyclohexanecarbonyl chloride 98%
Catalog # ACRO169051000
Supplier: Thermo Fisher Scientific
CAS Number: 2719-27-9
Jump to:
Product Details & Documents
Synonyms: Cyclohexanecarboxylic acid chloride, Cyclohexanecarboxylic chloride
- CAS Number:2719-27-9
- Formula:C₇H₁₁ClO
- MDL Number:MFCD00001456
- Molecular Weight:146.62 g/mol
- Boiling Pt:184 °C (1013 hPa)
- Melting Pt:29 °C
- Density:1.09 g/cm³
- Flash Pt:66 °C
- Storage Temperature:Ambient
- EINECS:220-322-7
- UN:3265
- ADR:8,II
Documents
ACRO16905
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless to straw yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4680 to 1.4700 (20°C, 589 nm)
- GC:≥97.5 %
- Titration RCOCl:≥97.5 %
- Titration RCOOH:≤2 %
- Free acid (HCl):Traces
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



