4-(Dimethylamino)benzaldehyde 99+%
Plastic bottle, 25 g
Supplier: Thermo Fisher Scientific
4-(Dimethylamino)benzaldehyde 99+%
Catalog # ACRO168760250
Supplier: Thermo Fisher Scientific
CAS Number: 100-10-7
Jump to:
Product Details & Documents
Synonyms: Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde
- CAS Number:100-10-7
- Formula:(CH₃)₂NC₆H₄CHO
- MDL Number:MFCD00003381
- Molecular Weight:149.19 g/mol
- Merck Index:13,03257
- Boiling Pt:176…177 °C (17 mmHg)
- Melting Pt:70…75 °C
- Density:1,0254 g/cm³ (99.9 °C)
- Flash Pt:164 °C
- Storage Temperature:Ambient
- EINECS:202-819-0
Specifications
Specification Test Results
- Appearance:Light yellow to light green Crystalline powder
- Infrared spectrum:Conforms
- Melting point:72 to 75 °C
- GC:≥99.0 %
Specifications
- Pack type:Plastic bottle
- Pk:25 g



