1,4-Dioxane 99+% for spectroscopy
Glass bottle, 2,5 L
: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide
- :123-91-1
- :C₄H₈O₂
- :MFCD00006571
- :88.11 g/mol
- :13,03330
- :101 °C (760 mmHg)
- :12 °C
- :1.034 g/cm³
- :Ambient
- :204-661-8
- :1165
- :3,II
- Appearance:Clear liquid
- Water:≤0.03 %
- GC:≥99.0 %
- UV:at 220 nm A: ≤1
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0005 %
- Pack type:Glass bottle
- Pk:2,5 L



