Cyclohexane 99+% for spectroscopy
Glass bottle, 2,5 L
Supplier: Thermo Fisher Scientific
Cyclohexane 99+% for spectroscopy
Catalog # ACRO167740025
Supplier: Thermo Fisher Scientific
CAS Number: 110-82-7
Jump to:
Product Details & Documents
Synonyms: Hexahydrobenzene, Hexamethylene
- CAS Number:110-82-7
- Formula:C₆H₁₂
- MDL Number:MFCD00003814
- Molecular Weight:84.16 g/mol
- Merck Index:13,02752
- Boiling Pt:81 °C (1013 hPa)
- Melting Pt:6.5 °C
- Density:0.77 g/cm³
- Flash Pt:–18 °C
- Storage Temperature:Ambient
- EINECS:203-806-2
- UN:1145
- ADR:3,II
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.01 %
- GC:≥99.0 %
- UV:at 210 nm A: ≤1
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0005 %
- Acidity:≤0.0002 meq/g
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



