Order Entry
Export
Orders LinkContactUsLinkComponent
Acetone 99+% for spectroscopy

Acetone 99+% for spectroscopy

Glass bottle, 500 mL
Catalog # ACRO167645000
Irritant
Flammable
   Safer Human and Environmental Health
Acetone 99+% for spectroscopy
Catalog # ACRO167645000
   Safer Human and Environmental Health
CAS Number:  67-64-1
Acetone 99+% for spectroscopy
Acetone 99+% for spectroscopy

Glass bottle, 500 mL

CAS Number:  67-64-1
Price
33,20 €
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56.2 °C (1013 hPa)
  • Melting Pt:–95 °C
  • Density:0.79 g/cm³
  • Flash Pt:–20 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.5 % (Coulometric)
  • GC:
    ≥99.0 %
  • UV:
    at 330 nm A: ≤1
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.001 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Cat. No.:
    ACRO167645000
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications