Order Entry
Export
Orders LinkContactUsLinkComponent
Capric acid 99%

Capric acid 99%

Glass bottle for solids, 500 g
Catalog # ACRO167275000
Irritant
Capric acid 99%
Catalog # ACRO167275000
CAS Number:  334-48-5
Capric acid 99%
Capric acid 99%

Glass bottle for solids, 500 g

CAS Number:  334-48-5
Price
142,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Decylic acid, Decanoic acid
  • CAS Number:334-48-5
  • Formula:C₁₀H₂₀O₂
  • MDL Number:MFCD00004441
  • Molecular Weight:172.27 g/mol
  • Merck Index:13,01764
  • Boiling Pt:268…270 °C (760 mmHg)
  • Melting Pt:30…32 °C
  • Density:0.89 g/cm³
  • Flash Pt:150 °C
  • Storage Temperature:Ambient
  • EINECS:206-376-4

Documents

Specifications

Specification Test Results

  • Appearance:
    Colorless or white to cream Crystals and/or chunks or fused solid or liquid
  • Infrared spectrum:
    Conforms
  • Melting point:
    29 to 34 °C
  • GC:
    ≥98.5 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications